missing translation for 'onlineSavingsMsg'
Learn More

Ammonium pentaborate octahydrate, 99.0%

Product Code. 11331649
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331649 500 g 500g
11321649 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331649 Supplier Thermo Scientific Alfa Aesar Supplier No. 012294.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12046-03-6 | (NH4)2B10O16·8H2O

Ammonium pentaborate octahydrate is used as a component of electrolyte solutions in electrolytic capacitors. It is also used as a flame retarder and in wood and textile industry. Further, it is used for brazing, welding, soldering fluxes and in some detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12046-03-6
MDL Number MFCD00135839

Specifications

Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0%
Molecular Formula (NH4)2B10O16·8H2O
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 544.29 (400.17 Anhydrous)
Chemical Name or Material Ammonium Pentaborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.