missing translation for 'onlineSavingsMsg'
Learn More

Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number 11493973
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
6009-70-7
C2H10N2O5
142.111
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
This item is not returnable. View return policy
6009-70-7
C2H10N2O5
142.111
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 70°C (decomposition)
Density 1.5 g/mL
Assay Percent Range >99%
Linear Formula (COONH4)2·H2O
Quantity 100 g
UN Number UN3288
Beilstein 3759830
Merck Index 14,537
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 142.11 (124.10Anhydrous)
Percent Purity 98%
Chemical Name or Material Ammonium oxalate monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'