missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium O,O'-diethyl dithiophosphate, typically 95%, Thermo Scientific Chemicals
52.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Diethyl dithiophosphate ammonium salt can be used in the extraction of arsenic. It is used as a source of the (C2H5O)2PS2- ligand in coordination chemistry and in analytical chemistry for determination of various ions. It can be obtained by the reaction of phosphorus pentasulfide with ethanol and ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 165°C to 175°C |
Color | White |
Odor | Unpleasant |
Linear Formula | (CH3CH2O)2P(S)SNH4 |
Quantity | 25 g |
Solubility Information | Soluble in water,methanol,ethanol,pyridine,acetone & ammonia. Insoluble in ethyl acetate,methyl chloride and chloroform. |
Formula Weight | 203.25 |
Percent Purity | 95% (typically) |
Physical Form | Crystalline |
Chemical Name or Material | Ammonium O,O'-diethyl dithiophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'