missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Nitrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Description
H4N2O3, CAS Number-6484-52-2, 500g, 210 deg.C, White, 80.04g/mol, 169 deg.C, 11425, Plastic powder jar, 4.5 to 6.0, Solid
Specifications
Boiling Point | 210°C |
Melting Point | 169°C |
Color | White |
pH | 4.5 to 6.0 |
Physical Form | Solid |
Quantity | 2.5 kg |
Formula Weight | 80.04g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Ammonium nitrate
- Warning
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'