missing translation for 'onlineSavingsMsg'
Learn More

Ammonium nitrate Analytical Reagent, Reag. ACS Reag. Ph. Eur., Solstice

Product Code. 15683230
Change view
Click to view available options
Quantity:
6 x 1 kg
Packaging:
Plastic Bottle
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15683230 6 x 1 kg Plastic Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15683230 Supplier Fluka Supplier No. 311146X1KG
  • ONLINE EXCLUSIVE PRICE
    £324.00
    £304.11 / Pack of 6
    Save £19.89
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥98%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6484-52-2
Physical Form Crystalline
Quantity 6 x 1 kg
Packaging Plastic Bottle
Molecular Formula H4N2O3
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES [NH4+].[O-][N+]([O-])=O
IUPAC Name ammonium nitrate
Molecular Weight (g/mol) 80.04
PubChem CID 22985
ChEBI CHEBI:63038
Percent Purity ≥98.0%
Grade ACS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.