missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
5g
50g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 5g |
Assay Percent Range | 100.00% |
Solubility Information | Highly soluble in water; soluble in alcohol and liquid ammonia. |
Formula Weight | 80.04 |
Odor | Odorless |
Appearance | Colorless crystalline |
Density | 1.72 g/cm3 |
Chemical Name or Material | Ammonium nitrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'