missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, Tetrahydrate, MP Biomedicals™

Product Code. 11442782 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442782 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442782 Supplier MP Biomedicals Supplier No. 0215249791
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ammonium Molybdate, Tetrahydrate, MP Biomedicals is an analgesic.

Specifications

CAS 12054-85-2
Physical Form Crystalline Powder
Quantity 1 kg
Molecular Formula (NH4)6Mo7O24.4H2O
Synonym Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate
Solubility Information Soluble in water (1g/mL).
Formula Weight 1235.86
Grade ACS
Chemical Name or Material Ammonium Molybdate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.