missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate tetrahydrate, Honeywell

Product Code. 15625510 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 15625510

Brand: Honeywell Research Chemicals 431346250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 12054-85-2
MDL Number MFCD00167059

Specifications

Quantity 250 g
Linear Formula (NH4)6Mo7O24 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 1235.86g/mol
Concentration or Composition (by Analyte or Components) 81 to 83% MoO3 basis, 99.98% (trace metals basis)
Density 2.498g/mL at 25°C
Chemical Name or Material Ammonium molybdate tetrahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.