missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate tetrahydrate, For ACS analysis, 81 to 83% MoO3, MP Biomedicals™

Product Code. 11432782 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432782 500 g 500g
11422782 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432782 Supplier MP Biomedicals Supplier No. 0215249790
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 12054-85-2
Molecular Formula (NH4)6 Mo7O24·4H2O
Synonym Molybdic acid ammonium salt tetrahydrate

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 81 to 83% MoO3
Density 2.87g/mL
Formula Weight 1235.86
Grade ACS
Chemical Name or Material Ammonium molybdate tetrahydrate, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.