missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate tetrahydrate, ACS Reagent, Solstice

Product Code. 15627670
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
Unit Size:
100g
1kg
2.5kg
500g
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15627670 100 g 100g
15657670 500 g 500g
15637670 1 kg 1kg
15647670 2.5 kg 2.5kg
15667670 6 x 500 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15627670 Supplier Honeywell-Fluka Supplier No. A7302100G
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

ACS Reagent, 81.0-83.0% MoO3O basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
MDL Number MFCD00167059

Specifications

Quantity 100 g
Linear Formula (NH4)6Mo7O24 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 1235.86g/mol
Grade ACS Reagent
Concentration or Composition (by Analyte or Components) 81 to 83% (MoO3 basis)
Density 2.498g/mL at 25°C
Chemical Name or Material Ammonium molybdate tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.