missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}

Product Code. 11341878
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341878 50 g 50g
11351878 100 g 100g
11321878 500 g 500g
11331878 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11341878 Supplier Thermo Scientific Alfa Aesar Supplier No. 011831.18
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

Ammonium molybdate (para) tetrahydrate, H24Mo7N6O24.4H2O, CAS Number-12054-85-2, Ammonium paramolybdate, 2kg, Powder, 81-83%, 2.498, 234-722-4, 1235.86 (1163.79 Anhydrous), ACS, H302-H315-H319-H335, P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c | CAS: 12054-85-2 | H24Mo7N6O24·4H2O

Ammonium molybdate (para) tetrahydrate, is used in photography,analytical lab reagent for phosphate,lead,arsenate determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24Mo7N6O24·4H2O
MDL Number MFCD00167059
Synonym Ammonium paramolybdate

Specifications

Color Powder
Melting Point ∽190°C (decomposition)
Assay Percent Range 81-83%
Density 2.498 g/cm3
Solubility Information Soluble in water (1:2 ratio). Insoluble in alcohol
Formula Weight 1235.86 (1163.79 Anhydrous)
Grade ACS
Odor Odorless
Quantity 50 g
Physical Form Powder
Chemical Name or Material Ammonium molybdate (para) tetrahydrate, (para) tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.