missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate (di), Mo 56.5%

Product Code. 11316628
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316628 50 g 50g
11336628 250 g 250g
11326628 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11316628 Supplier Thermo Scientific Alfa Aesar Supplier No. 012215.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27546-07-2 | H2MoO4 | 161.96 g/mol

Ammonium molybdate (di) is used as pigments, agricultural fertilizer, color lakes and fabric fire retardant agent. It is also used as a raw material for the production of molybdenum wire, ceramic glazes and other molybdenum compounds. Further, it is used in photography, ceramics and paints. It is also used reagent for analysis as well as in the determination of phosphorus in analytical chemistry. In addition, it is used as a catalyst in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27546-07-2
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00010882
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym Ammonium dimolybdate

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 339.95
Concentration or Composition (by Analyte or Components) Mo 56.5%
Chemical Name or Material Ammonium molybdate (di)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.