missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate (di), 10% w/v aq. soln.

Product Code. 11373677
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373677 1 L 1 L
11485020 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373677 Supplier Thermo Scientific Alfa Aesar Supplier No. 035579.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27546-07-2
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00010882
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L

Specifications

Concentration 10% w/v aq. soln.
Formula Weight 339.95
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Ammonium molybdate (di)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.