missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Tetrahydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31749658
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31749658 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749658 Supplier Ricca Chemical Company Supplier No. RDCA0450500B1
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 12054-85-2
Color Green-Yellow to White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Molecular Weight (g/mol) 1235.86 g/mol
Percent Purity 81.0 to 83.0%
Grade ACS
Concentration or Composition (by Analyte or Components) 4-Hydrate
Chemical Name or Material Ammonium Molybdate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.