missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Metavanadate, (White to light yellow powder),MP Biomedicals

Product Code. 11465062 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
11465062 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 11465062

Brand: MP Biomedicals 0219462480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7803-55-6
Color White to Yellow
Physical Form Powder
Quantity 100 g
Molecular Formula H4NO3V
MDL Number MFCD00011430
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILES N.O[V](=O)=O
IUPAC Name vanadiumoylol amine
Molecular Weight (g/mol) 116.98
PubChem CID 516859
Formula Weight 117
Chemical Name or Material Ammonium Metavanadate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.