missing translation for 'onlineSavingsMsg'
Learn More

Ammonium metavanadate, Puriss. p.a., Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Product Code. 15603290 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
50 kg
Unit Size:
100g
1kg
50kg
Product Code. Quantity unitSize
15603290 100 g 100g
15613290 1 kg 1kg
15623290 50 kg 50kg
3 options
This item is not returnable. View return policy

Product Code. 15603290

Brand: Honeywell-Fluka 31153100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

Quantity 100 g
Linear Formula NH4VO3
UN Number 2859
Formula Weight 116.98g/mol
Percent Purity ≥99.5%
Grade puriss. p.a.
Packaging Plastic Bottle
Density 2.32g/cm3at 25°C
Chemical Name or Material Ammonium Metavanadate, Reag. Ph. Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.