missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Metavanadate, ACS Reagent, ≥99%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
Unit Size:
250g
50g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 200°C |
| Color | Yellow |
| pH | 7 |
| Quantity | 50 g |
| Linear Formula | NH4VO3 |
| UN Number | UN2859 |
| Solubility Information | In water 5.1g/L at 20°C |
| Formula Weight | 116.98g/mol |
| Percent Purity | ≥99% |
| Concentration or Composition (by Analyte or Components) | 5.1g/L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction