missing translation for 'onlineSavingsMsg'
Learn More

Ammonium L-(+)-tartrate, 98%

Product Code. 11343387
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343387 250 g 250g
11333387 50 g 50g
11323387 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343387 Supplier Thermo Scientific Alfa Aesar Supplier No. 017658.30
 Restricted Item
In Stock
Add to basket
Edge
Aggiungi al carrello
Request Bulk or Custom Formats
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol

Ammonium L-(+)-tartrate finds application in the textile and pharmaceutical industries. It is also used to study protein structural analysis, biochemicals and reagents, biological buffers, chelators and proteomics. It acts as a nitrogen source in biological applications and cell culture media. Further, it is used in the study of crystal optimization research and preliminary diffraction data analysis of the Smad1 MH1 domain bonded to a palindromic SBE DNA element.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid diamine
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifica

Physical Form Crystalline
Quantity 250 g
Beilstein 6120352
Solubility Information Soluble in water and alcohol.
Formula Weight 184.15
Sensitivity Hygroscopic
Density 1.6
Chemical Name or Material Ammonium-L-tartrate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.