missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(III) Sulfate Dodecahydrate, Certified AR for analysis, Fisher Chemical™

Product Code. p-7072818 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10569180

Brand: Fisher Chemical A/4760/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H4FeNO8S2.12H2O, CAS Number-7783-83-7, 500g, 230 deg.C, Purple, 482.19g/mol, 39 deg.C, 150004, Plastic powder jar, 2.5, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.18
MDL Number 150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
IUPAC Name iron(3+) ammonium dodecahydrate disulfate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Boiling Point 230°C
Melting Point 39°C
Color Purple
pH 2.5
Physical Form Solid
Quantity 500 g
Formula Weight 482.19g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.