missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, 98+%

Product Code. 11453137
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453137 2500 g 2500g
11443137 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453137 Supplier Thermo Scientific Alfa Aesar Supplier No. A16105.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-83-7 | FeH28NO20S2 | 482.176 g/mol

Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

Solubility
Soluble in water (1240 mg/ml at 25°C) and 1% (v/v) aqueous HCl (100 mg/ml). Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Melting Point 39°C to 41°C
Quantity 2500 g
Linear Formula NH4Fe(SO4)2·12H2O
Merck Index 14,521
Solubility Information Soluble in water (1240mg/ml at 25°C) and 1% (v/v) aqueous HCl (100mg/ml). Insoluble in alcohol.
Formula Weight 482.18 (266.00 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Density 1.71
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate
compliance-icons
Product Identifier
  • Ammonium iron(III) sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.