missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iron(III) oxalate trihydrate, 98%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
50g
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Crystalline Powder |
| Quantity | 5 kg |
| Assay Percent Range | 98% |
| Linear Formula | (NH4)3Fe(C2O4)3·3H2O |
| Merck Index | 10,533 |
| Formula Weight | 428.07 |
| Percent Purity | 98% |
| Grade | Pure |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
Product Identifier
- Ammonium iron(III) oxalate trihydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction