missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) hexacyanoferrate(II) hydrate, tech., Thermo Scientific Chemicals

Catalog Number 11444987
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
25869-00-5
C6H4Fe2N7
285.837
MFCD01726733
ILZWGESBVHGTRX-UHFFFAOYSA-O
ammonium iron iii hexacyanoferrate ii, iron 3+ ammonium ferrocyanide, azanium;iron 2+ ;iron 3+ ;hexacyanide, ammonium iron iii hexacyanoferrate ii hydrate, ammonium iron iii hexacyanoferrate ii hydrate, tech., ammonium iron iii hexacyanoferrate ii giese salt
23620040
azanium;iron(2+);iron(3+);hexacyanide
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[Fe+2].[Fe+3]
This item is not returnable. View return policy
25869-00-5
C6H4Fe2N7
285.837
MFCD01726733
ILZWGESBVHGTRX-UHFFFAOYSA-O
ammonium iron iii hexacyanoferrate ii, iron 3+ ammonium ferrocyanide, azanium;iron 2+ ;iron 3+ ;hexacyanide, ammonium iron iii hexacyanoferrate ii hydrate, ammonium iron iii hexacyanoferrate ii hydrate, tech., ammonium iron iii hexacyanoferrate ii giese salt
23620040
azanium;iron(2+);iron(3+);hexacyanide
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[Fe+2].[Fe+3]

It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

Solubility
Soluble in water and mineral acids. Insoluble in alkali.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 25 g
Merck Index 14,7910
Solubility Information Soluble in water and mineral acids. Insoluble in alkali.
Formula Weight 285.83 (Anhydrous)
Grade Technical
Chemical Name or Material Ammonium iron(III) hexacyanoferrate(II) hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'