missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron (III) citrate, J.T. Baker™

Product Code. 10666102 Shop All J.T.Baker Products
Click to view available options
:
1 kg
This item is not returnable. View return policy

Product Code. 10666102

Brand: JT Baker 1110.1000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratory Reagent

Specifications

CAS 1185-57-5
Quantity 1 kg
Assay Percent Range 14.5 to 16.0% (as Fe)
Molecular Formula C6H8FeNO7
MDL Number MFCD00013099
Synonym Ammonium iron (III) citrate , Ferric Ammonium Citrate , Ammonium ferric citrate
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-K
SMILES N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
IUPAC Name iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
Molecular Weight (g/mol) 261.98
Packaging Glass bottle for solids
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.