missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(Iii) Citrate, Honeywell™

Product Code. 15620890 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
500 g
1 kg
Unit Size:
1kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15620890 500 g 500g
15630890 50 g 50g
15610890 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15620890 Supplier Honeywell Research Chemicals Supplier No. 09713500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purum p.a., 14.5-16% Fe basis (RT), powder, green

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Green
Physical Form Powder
Quantity 500 g
Linear Formula C6H8O7 ·xFe3+ · yNH3
Grade purum p.a.
Concentration or Composition (by Analyte or Components) 14.5 to 16% Fe basis (RT)
Chemical Name or Material Ammonium Iron(Iii) Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.