missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) citrate, brown

Product Code. 11453504
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11453504

Brand: Thermo Scientific Alfa Aesar A11199.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1185-57-5 | C6H11FeNO7+3 | 264.999 g/mol

Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

Solubility
Soluble in water.1200 g/L (20°C)

Notes
Hygroscopic. Desiccate at room temperature. Store in the dark. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
MDL Number MFCD00013099
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Brown
Quantity 1000 g
Merck Index 14,4017
Solubility Information Soluble in water.1200g/L (20°C)
Odor Weak
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Ammonium iron (III) citrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.