missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) citrate, 16.5 to 18.5% Fe

Product Code. 10424395 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10424395 2.5 kg Plastic bottle 2.5kg
10762505 250 g Plastic bottle 250g
10104700 1 kg Plastic bottle 1kg
11985061 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10424395 Supplier Thermo Scientific Acros Supplier No. 211840025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1185-57-5 | C6H11FeNO7 | 265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7
Molecular Weight (g/mol) 265
MDL Number MFCD00013099
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Brown to Brown-Yellow
Physical Form Powder
Quantity 2.5 kg
Merck Index 15, 4044
Solubility Information Solubility in water: 1200g/L (20 c). Other solubilities: practically insoluble in alcohol
Formula Weight 265
Percent Purity 16.5 to 18.5% (Fe)
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Ammonium iron(III) citrate, 16.5 to 18.5% Fe

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.