missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(II) Sulfate Solution 0.025M, Standard Solution ready to use for volumetric analysis, Fisher Chemical

Product Code. 10743991 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10743991 2.5 L Amber glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10743991 Supplier Fisher Chemical Supplier No. J/1100/17

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7783-85-9
Physical Form Liquid
Quantity 2.5 L
Molecular Formula FeH20N2O14S2
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
Molecular Weight (g/mol) 392.13
Packaging Amber glass bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.