missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(II) Sulfate Hexahydrate (Mohr’s Salt), TRC
Specifications
Specifications
| Chemical Name or Material | Ammonium Iron(II) Sulfate Hexahydrate (Mohr's Salt) |
| CAS | 7783-85-9 |
| Quantity | 50 g |
| Molecular Formula | Fe . 2 H O4 S . 6 H2 O . 2[N H3] |
| Synonym | Ammonium Ferrous Sulfate Hexahydrate, Ammonium Iron Disulfate Hexahydrate ((NH4)2Fe(SO4)2.6H2O), Ammonium Iron Sulfate ((NH4)2Fe(SO4)2) Hexahydrate, Diammonium Iron Disulfate Hexahydrate, Diammonium Iron Sulfate ((NH4)2Fe(SO4)2) Hexahydrate, Diammonium iron(2+) Disulfate Hexahydrate, Diammonium iron(II) Disulfate Hexahydrate, Diammonium iron(II) Sulfate Hexahydrate, Ferrous Ammonium Sulfate [FeSO4.(NH4)2SO4.6H2O], Ferrous Ammonium Sulfate Hexahydrate, Iron Ammonium Sulfate Hydrate (FeSO4(H4)2SO4.6H2O) |
| InChI Formula | InChI=1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2 |
| SMILES | N.N.O.O.O.O.O.O.[Fe+2].OS(=O)(=O)[O-].OS(=O)(=O)[O-] |
| IUPAC Name | azane;hydrogen sulfate;iron(2+);hexahydrate |
| Molecular Weight (g/mol) | 392.139 |
| Formula Weight | 391.971 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?