missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(II) Sulfate Hexahydrate, ≥98%, Reagent Grade, Honeywell™
Reagent Grade,≥98%
£45.25 - £272.18
Chemical Identifiers
CAS | 7783-85-9 |
---|---|
Molecular Formula | FeH20N2O14S2 |
Molecular Weight (g/mol) | 392.13 |
MDL Number | MFCD00150530 |
InChI Key | MQLVWQSVRZVNIP-UHFFFAOYSA-L |
IUPAC Name | λ²-iron(2+) diammonium hexahydrate disulfate |
SMILES | [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15627870
|
Honeywell Research Chemicals
F3754-500G |
500 g | Poly bottle | ||||||
15617870
|
Honeywell Research Chemicals
F3754-1KG |
1 kg | Poly bottle | ||||||
15637870
|
Honeywell Research Chemicals
F3754-5KG |
5 kg | Poly bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7783-85-9 | |
392.13 | |
MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
FeH20N2O14S2 | |
MFCD00150530 | |
λ²-iron(2+) diammonium hexahydrate disulfate |
Specifications
100°C | |
Blue Green | |
FeH20N2O14S2 | |
MFCD00150530 | |
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
392.13 | |
≥98% | |
1.86g/cu.cm at 20°C |
7783-85-9 | |
Solid | |
(NH4)2Fe(SO4)2 · 6H2O | |
MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
λ²-iron(2+) diammonium hexahydrate disulfate | |
392.14g/mol | |
Odorless | |
Ammonium iron(II) sulfate hexahydrate |
Safety and Handling
missing translation for 'rtecsNumber' : 233-151-8