missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodide, 99.995% (metals basis)

Product Code. 11333778
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333778 25 g 25g
11323778 5 g 5g
11343778 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333778 Supplier Thermo Scientific Alfa Aesar Supplier No. 040255.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Light Sensitive and Hygroscopic | CAS: 12027-06-4 | H4IN | 144.94 g/mol

It is used in photographic chemicals and some medications. Ammonium iodide and hydrogen peroxide plays an essential role for the eco-friendly oxyiodination of aromatic compounds. Ammonium Iodide and DMF is also useful for the copper-mediated selective cyanation reaction of Indoles and 2-Phenylpyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Melting Point 551° (sublimation)
Color White
Physical Form Granules
Quantity 25 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Soluble in water and ethanol.
Formula Weight 144.94
Odor Odorless
Refractive Index 1.7031
Sensitivity Air and light sensitive; Hygroscopic
Appearance White granules
Vapor Pressure 1.3 hPa (1mm Hg)
Density 2.514 g/cm3
Chemical Name or Material Ammonium Iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.