missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407222
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
12027-06-4
H4IN
144.94
MFCD00011424
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]
This item is not returnable. View return policy
12027-06-4
H4IN
144.94
MFCD00011424
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]

Light Sensitive and Hygroscopic

It is used in photographic chemicals and some medications. Ammonium iodide and hydrogen peroxide plays an essential role for the eco-friendly oxyiodination of aromatic compounds. Ammonium Iodide and DMF is also useful for the copper-mediated selective cyanation reaction of Indoles and 2-Phenylpyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 551°C (sublimation)
Color White to Yellow
Quantity 5 g
Linear Formula NH4I
Merck Index 14,529
Solubility Information Soluble in water and ethanol.
Formula Weight 144.94
Percent Purity 99.995%
Odor Odorless
Refractive Index 1.7031
Sensitivity Air and light sensitive; Hygroscopic
Density 2.514
Chemical Name or Material Ammonium iodide, (metals basis)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'