missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodate, for analysis, Thermo Scientific™

Product Code. 10330184 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Plastic bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10330184 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10330184 Supplier Thermo Scientific Acros Supplier No. 390550500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-09-8
Molecular Weight (g/mol) 192.94
MDL Number MFCD00049684
InChI Key ZRDJERPXCFOFCP-UHFFFAOYSA-N
Synonym ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
PubChem CID 166805
IUPAC Name azanium;iodate
SMILES [NH4+].[O-]I(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 50 g
Molecular Formula H4INO3
Solubility Information Solubility in water: 2g/L (15°C)
Formula Weight 192.94
Percent Purity ≥99.0%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Ammonium iodate
compliance-icons compliance-icons
Product Identifier
  • Ammonium iodate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.