missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodate, 99%

Product Code. 11372478
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372478 50 g 50g
11362478 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372478 Supplier Thermo Scientific Alfa Aesar Supplier No. 014531.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13446-09-8 | H4INO3 | 192.94 g/mol

Ammonium iodate is used as an oxidation reagent. Ammonium Iodate-18O3 is the labeled analog of ammonium iodate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-09-8
Molecular Formula H4INO3
Molecular Weight (g/mol) 192.94
MDL Number MFCD00049684
InChI Key ZRDJERPXCFOFCP-UHFFFAOYSA-N
Synonym ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
PubChem CID 166805
IUPAC Name azanium;iodate
SMILES [NH4+].[O-]I(=O)=O

Specifications

Melting Point 150°C
Color White
Physical Form Crystalline
Quantity 50 g
Linear Formula NH4IO3
UN Number UN1479
Solubility Information Soluble in water 20.6g/L @ 20°C
Formula Weight 192.94
Percent Purity 99%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.309 g/mL
Chemical Name or Material Ammonium iodate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.