Learn More
Ammonium iodate, 99%
Description
Ammonium iodate is used as an oxidation reagent. Ammonium Iodate-18O3 is the labeled analog of ammonium iodate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 150°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Linear Formula | NH4IO3 |
| UN Number | UN1479 |
| Solubility Information | Soluble in water 20.6g/L @ 20°C |
| Formula Weight | 192.94 |
| Percent Purity | 99% |
| Odor | Odorless |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.