missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide Solution, Honeywell™

Product Code. 15691570 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15691570 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15691570 Supplier Honeywell-Fluka Supplier No. 17837100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 1336-21-6
Molecular Formula H5NO
Linear Formula NH4OH
Concentration approx.10% in water
MDL Number MFCD00066650
UN Number UN2672
Beilstein 3587154
Density 0.9g/mL at 25°C
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 100 mL
SMILES N.O
IUPAC Name amine hydrate
Explosion Limit 0.27
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.05g/mol
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.