missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide solution, Eluent additive for LC-MS, ≥25% in H2O, Solstice

Product Code. 15655540
Change view
Click to view available options
Quantity:
100 mL
10 x 1 mL
Packaging:
Ampoule
Glass Bottle
Polybottle
Unit Size:
100mL
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15655540 100 mL Glass Bottle 100mL
15628410 10 x 1 mL Ampoule Pack of 10
15645540 100 mL Polybottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15655540 Supplier Honeywell-Fluka Supplier No. 44273100MLGL
  • ONLINE EXCLUSIVE PRICE
    £125.00
    £62.11 / 100mL
    Save £62.89
    50% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

Eluent additive for LC-MS,≥25% in H2O

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Boiling Point 24°C
CAS Min % ≥70.0000%, ≥20.0000%
CAS Max % <90.0000%, <30.0000%
Linear Formula NH4OH
UN Number UN2672
Beilstein 3587154
Density 0.9 g/mL at 25 °C(lit.)
Quantity 100 mL
IUPAC Name amine hydrate
Explosion Limit 0.27
Formula Weight 35.05
Percent Purity >25%
Grade LC-MS
Packaging Glass Bottle
Chemical Name or Material Ammonium hydroxide solution, Eluent additive for LC-MS, ≥25% in H2O
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonia, aqueous solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.