missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, ACS, 28.0-30.0% NH{3}, Thermo Scientific Chemicals

Catalog Number 11327598
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O

Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silve, and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Assay Percent Range 28.0-30.0% NH3
Density 0.90 g/cm3
Solubility Information Miscible with water.
Quantity 250 g
IUPAC Name amine hydrate
Formula Weight 35.05
Grade ACS
Odor Ammonia-like
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'