missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Product Code. 11379299
Change view
Click to view available options
Quantity:
1 Unit
6 Units
Unit Size:
1 unit
6 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379299 6 Units 6 units
11369299 1 Unit 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379299 Supplier Thermo Scientific Alfa Aesar Supplier No. 088576.LS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1336-21-6 | H5NO | 35.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Odor Ammonia-like
Assay Percent Range 0.1N
Physical Form Liquid
Color Colorless
Quantity 6 Units
Formula Weight 35.05
Chemical Name or Material Ammonium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.