missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)

Product Code. 11366329
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366329 1000 mL 1000mL
11399119 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366329 Supplier Thermo Scientific Alfa Aesar Supplier No. 087903.AU
  • ONLINE EXCLUSIVE PRICE
    £443.00
    £379.65 / 1000mL
    Save £63.35
    14% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1336-21-6 | H5NO | 35.05 g/mol

Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silver and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Spécification

Color Colorless
Assay Percent Range 99.99% (metals basis)
Density 0.91
Solubility Information Miscible with water.
Quantity 1000 mL
IUPAC Name azanium;hydroxide
Formula Weight 35.05
Odor Ammonia-like
Vapor Pressure 362mm Hg at 20°C
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.