missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen sulfate, extra pure

Product Code. 11914261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11914261 1 kg 1kg
11924261 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11914261 Supplier Thermo Scientific Acros Supplier No. 445910010
  • ONLINE EXCLUSIVE PRICE
    £59.55
    £57.80 / 1kg
    Save £1.75
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7803-63-6 | H5NO4S | 115.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-63-6
Molecular Formula H5NO4S
Molecular Weight (g/mol) 115.10
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
PubChem CID 24655
SMILES [NH4+].OS([O-])(=O)=O

Specifications

Boiling Point 350.0°C
Melting Point 147.0°C
Quantity 1 kg
Assay Percent Range Extra pure
Linear Formula NH4HSO4
Merck Index 15,497
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: practically insoluble in alcohol, acetone and, pyridine
IUPAC Name ammonium hydrogen sulfate
Formula Weight 115.11
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Ammonium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.