missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydrogen Sulfate, Extra Pure, SLR, Fisher Chemical

Product Code. 10458780 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10458780 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10458780 Supplier Fisher Chemical Supplier No. A/5400/53
  • ONLINE EXCLUSIVE PRICE
    £220.54
    £194.00 / 500g
    Save £26.54
    12% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 350°C
Melting Point 147°C
CAS 7803-63-6
Color White
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula H5NO4S
MDL Number 51073
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
SMILES [NH4+].OS([O-])(=O)=O
IUPAC Name ammonium hydrogen sulfate
Molecular Weight (g/mol) 115.10
Formula Weight 115.11g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.