missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen sulfate, 99%, for analysis

Product Code. 10370734 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10370734 250 g Glass bottle 250g
10065073 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10370734 Supplier Thermo Scientific Acros Supplier No. 389412500
  • ONLINE EXCLUSIVE PRICE
    £22.90
    £22.20 / 250g
    Save £0.70
    3% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7803-63-6 | H5NO4S | 115.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-63-6
Molecular Formula H5NO4S
Molecular Weight (g/mol) 115.10
MDL Number MFCD00051073
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
PubChem CID 24655
SMILES [NH4+].OS([O-])(=O)=O

Specifications

Boiling Point 350.0°C
Melting Point 147.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99%
Linear Formula NH4HSO4
Merck Index 15, 497
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: practically insoluble in alcohol, acetone and, pyridine
IUPAC Name azanium;hydrogen sulfate
Formula Weight 115.11
Percent Purity 99%
Grade Analytical
Packaging Glass bottle
Residue after Ignition 0.01% max. (600°C)
Chemical Name or Material Ammonium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.