missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen sulfate, 99.9% (metals basis)

Product Code. 11342438
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342438 500 g 500g
11322438 25 g 25g
11332438 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342438 Supplier Thermo Scientific Alfa Aesar Supplier No. 014485.36
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7803-63-6 | H5NO4S | 115.10 g/mol

Ammonium hydrogen sulfate is used as pharmaceutical intermediates and chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-63-6
Molecular Formula H5NO4S
Molecular Weight (g/mol) 115.10
MDL Number MFCD00051073
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
PubChem CID 24655
SMILES [NH4+].OS([O-])(=O)=O

Specifications

Melting Point 146.9°C
Physical Form Lump
Quantity 500 g
UN Number UN2506
Solubility Information Soluble in water (∽115.1mg/ml at 20°C). Insoluble in acetone,alcohol,and pyridine.
IUPAC Name azanium;hydrogen sulfate
Formula Weight 115.1
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.