missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen phosphate, ACS, 98.0% min

Product Code. 11387337
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387337 250 g 250g
11397337 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387337 Supplier Thermo Scientific Alfa Aesar Supplier No. 011597.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol

Ammonium hydrogen phosphate is commonly used as a fertilizer. It can be used as fire retardant, in fireproofing textiles, paper and wood. It finds applications in vegetable fibers, impregnating lamp wicks, soldering metals, purifying sugar, in yeast cultures and corrosion inhibitors. It is used in the treatment of carbonate stones used in architectural, cultural heritage and optical materials. It is also used as a flux for soldering copper, tin and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name diazanium;hydrogen phosphate
SMILES N.N.OP(O)(O)=O

Specifications

Melting Point 155°C (decomposition)
Color Colorless
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 98% min.
Solubility Information Soluble in water. Insoluble in alcohol,acetone and liquid ammonia.
Formula Weight 132.06
Grade ACS
Density 1.619
Chemical Name or Material Ammonium hydrogen phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.