missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals

Product Code. 11396968
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396968 2 kg 2kg
11386968 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11396968 Supplier Thermo Scientific Alfa Aesar Supplier No. 014660.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Ammonium hydrogen fluoride is used as a chemical reagent in analysis and an antiseptic. It is also used in ceramics, electroplating, component of glass etchants as well as food processing equipment disinfectant. It acts as an intermediate in the production of hydrofluoric acid from hexafluorosilicic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium bifluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 238°C
Melting Point 120°C to 122°C
pH 2 to 3
Physical Form Flakes
Quantity 2 kg
UN Number UN1727
Merck Index 14,498
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 57.04
Percent Purity 95%
Grade Technical
Odor Ammonia-like
Sensitivity Hygroscopic
Density 1.500 g/mL
Chemical Name or Material Ammonium hydrogen fluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium hydrogen fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium bifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.