missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals

Product Code. 11324006
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324006 100 g 100g
11334006 500 g 500g
11344006 2500 g 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11324006 Supplier Thermo Scientific Alfa Aesar Supplier No. L17237.22
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1341-49-7 | Molecular Formula: F2H5N | Molecular Weight: 57.04 g/mol

Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium bifluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 238°C (decomposition)
Melting Point 120°C to 122°C
pH 2 to 3
Quantity 100 g
Odor Ammonia-like
Sensitivity Hygroscopic
Density 1.5
Linear Formula (NH4)HF2
UN Number UN1727
Merck Index 14,498
Solubility Information Soluble in water.
Formula Weight 57.04
Percent Purity 95%
Assay remainder mainly ammonium fluoride
Chemical Name or Material Ammonium hydrogen fluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium hydrogen fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium bifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.