missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydrogen Difluoride, Technical, Fisher Chemical

Product Code. 10599560 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
3 kg
Unit Size:
3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10599560 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10599560 Supplier Fisher Chemical Supplier No. A/5240/63
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Boiling Point 230°C
Melting Point 125°C
CAS 1341-49-7
Color White
pH 3.5
Physical Form Solid
Quantity 3 kg
Molecular Formula F2H5N
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILES N.F.F
IUPAC Name ammonium hydrogen difluoride
Molecular Weight (g/mol) 57.04
Formula Weight 57.04g/mol
Packaging Plastic powder jar
Vapor Pressure 1 hPa at 20°C
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium hydrogen difluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.