Learn More
Ammonium hydrogen carbonate, 99.0%, Thermo Scientific Chemicals
Description
In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition.Ammonium hydrogen carbonate is used as a reagent in chemical industries. It is an active component used in the manufacturing of fire-extinguishing compounds, pharmaceuticals, dyes, pigments and fertilizers. It finds application in the plastic, ceramics, leather and rubber industries. It is also used in cooling baths, scale and gypsum removing agents, leavening agent for baked goods and a defatting agent for textiles. Further, it acts as a buffer used in the purification of chemicals. In addition to this, it is involved in the gel digestion of proteins by trypsin and mass spectrometric analysis of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments
- Also used for compost treatment to accelerate decomposition
Specifications
Melting Point | ∼60°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | (NH4)HCO3 |
Beilstein | 4329606 |
Merck Index | 14,497 |
Solubility Information | Soluble in cold water but decomposed by hot water; Insoluble in alcohol and acetone. |
Formula Weight | 79.06 |
Percent Purity | 99.0% |
Show More |
RUO – Research Use Only