missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorosilicate, 99.999%, (trace metal basis)

Product Code. 10391230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10391230 250 g Glass bottle 250g
10352500 10 g Glass bottle 10g
10312550 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10391230 Supplier Thermo Scientific Acros Supplier No. 194002500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16919-19-0 | H8F6N2Si | 178.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-19-0
Molecular Formula H8F6N2Si
Molecular Weight (g/mol) 178.15
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
Synonym ammonium hexafluorosilicate, ammonium fluosilicate, ammonium fluorosilicate, ammonium silicofluoride, cryptohalite, caswell no. 043, diammonium fluosilicate, diammonium fluorosilicate, ammonium silicon fluoride, diammonium hexafluorosilicate
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 250 g
Assay Percent Range 99.999%
Linear Formula (NH4)2SiF6
Merck Index 15,524
Solubility Information Solubility in water: 18.6 g/100mL (17°C).
Formula Weight 178.15
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 2.01
Density 2.0100g/mL
Chemical Name or Material Ammonium hexafluorosilicate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.