missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorosilicate, 99.999%, (trace metal basis)

Product Code. 10312550 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10312550

Brand: Thermo Scientific Acros 194000500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16919-19-0 | H8F6N2Si | 178.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-19-0
Molecular Formula H8F6N2Si
Molecular Weight (g/mol) 178.15
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
Synonym ammonium hexafluorosilicate, ammonium fluosilicate, ammonium fluorosilicate, ammonium silicofluoride, cryptohalite, caswell no. 043, diammonium fluosilicate, diammonium fluorosilicate, ammonium silicon fluoride, diammonium hexafluorosilicate
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 50 g
Assay Percent Range 99.999%
Linear Formula (NH4)2SiF6
Merck Index 15,524
Solubility Information Solubility in water: 18.6 g/100mL (17°C).
Formula Weight 178.15
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 2.01
Density 2.0100g/mL
Chemical Name or Material Ammonium hexafluorosilicate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.